Learn More
Youngstown Barrel & Drum Plastic Open-Head Drums Available on GSA/VA Contract for Federal Government customers only.
High-density polyethylene containers for transport of hazardous solids or liquids
Manufacturer:  Youngstown Barrel & Drum 1713
 View more versions of this product
Catalog No. 19-120-002
Add to cart
Description
Description
  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
UN 3H2/Y90/S
4.33cm
50
Black lid
3.34 lb.
10 gal.
Blue, Black
11 in.
For transport of hazardous solids or liquids
Open
“Euro” Style Drum
1.52kg
37.84L
Plastic