Learn More
Youngstown Barrel & Drum Plastic Open-Head Drums - PROMO
High-density polyethylene containers for transport of hazardous solids
Manufacturer:  Youngstown Barrel & Drum 1778PMO
Catalog No. 19120008PMO
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking
Specifications
Specifications
“Euro” Style Drum
1.36kg
30.28L
Plastic
11 in.
For transport of hazardous solids or liquids
Open
47.62cm
3 lb.
8 gal.
Blue, Black
UN 3H2/Y51/S
4.33cm
50
18.75 in.
Black lid