Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Youngstown Barrel & Drum Plastic Open-Head Drums: PROMO

High-density polyethylene containers for transport of hazardous solids or liquids

Manufacturer:  Youngstown Barrel & Drum 1713PMO

Catalog No. 19120002PMO

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
Specifications

Specifications

“Euro” Style Drum
1.52kg
37.84L
Plastic
11 in.
For transport of hazardous solids or liquids
Open
3.34 lb.
10 gal.
Blue, Black
UN 3H2/Y90/S
4.33cm
50
Black lid
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit