Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Open-Head Drums

High-density polyethylene containers for transport of hazardous solids or liquids

Manufacturer:  Youngstown Barrel & Drum Company 1546

 View more versions of this product

Catalog No. 18-120-232

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking

Description & Specifications

Specifications

Drum
73.91cm
30 gal.
Lever lock ring
44.45cm
Plastic
50
32.18 lb.
30 gal., 113.56L
29.1 in.
113.56L
UN 1H2/Y180/S
17.5 in.
Blue
For transport of hazardous solids or liquids
14.6kg
Open
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit