Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Open-Head Drums Available on GSA/VA Contract for Federal Government customers only.

High-density polyethylene containers for transport of hazardous solids or liquids

Manufacturer:  Youngstown Barrel & Drum Company 1720

 View more versions of this product

Catalog No. 18-940


  Check Availability
Add to cart

Specifications

Description

  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking

Description & Specifications

Specifications

Drum
63.5cm
14 gal.
Side lever lock ring
34.29cm
Plastic
50
18.07 lb.
Open
25 in.
52.99L
UN 1H2/Y100/S and X65/S
13.5 in.
Blue
For transport of hazardous solids or liquids
8.2kg
Plain cover
14 gal., 52.99L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit