Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Open-Head Drums
SDP

High-density polyethylene containers for transport of hazardous solids or liquids

Supplier:  Youngstown Barrel & Drum 1748

 View more versions of this product

Catalog No. 18-120-570


Add to Cart

Description

Description

  • Made of chemical-resistant high-density polyethylene
  • Open-head drums for transport of hazardous solids
  • 19-120-008 and 19-120-004 feature flat sides for easier storage and stacking

Specifications

Specifications

UN 1H2/Y54/S
Screw Lid
40.5 L
For Transport of Hazardous Solids or Liquids
50
Screw Lid
52.73 cm
35.56 cm
6.8 Kg
Drum
10.7 gal.
White
Plastic
Open
20.76 in.
14 in.
14.99 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit