Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Tight Head Drums Available on GSA/VA Contract for Federal Government customers only.

High-density polyethylene containers for transport of hazardous solids or liquids

Manufacturer:  Youngstown Barrel & Drum Company 1419

 View more versions of this product

Catalog No. 18-120-237


  Check Availability
Add to cart

Specifications

Description

  • Made of chemical-resistant high-density polyethylene
  • Tight-head drums for transport of hazardous liquids

Description & Specifications

Specifications

Plastic Tight Head Drums
89.4cm
55 gal.
59.18cm
Translucent
For transport of hazardous solids or liquids
8.25kg
Fittings: 2 in. BTR and 2 in. NPS
55 gal., 208.19L
35.2 in.
208.19L
UN 1H1/Y1.9/150
23.3 in.
Plastic
50
18.2 lb.
Tight
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit