Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Plastic Tight Head Drums - PROMO
High-density polyethylene containers for transport of hazardous solids or liquids
Supplier: Youngstown Barrel & Drum 1419PMO/PROMO OVER
Description
- Made of chemical-resistant high-density polyethylene
- Tight-head drums for transport of hazardous liquids
Specifications
UN 1H1/Y1.9/150 | |
55 gal. | |
Translucent | |
Plastic | |
Tight | |
35.2 in. | |
23.3 in. | |
18.2 lb. |
Plastic Tight Head Drums | |
208.19 L | |
For Transport of Hazardous Solids or Liquids | |
50 | |
Fittings: 2 in. BTR and 2 in. NPS | |
89.4 cm | |
59.18 mm | |
8.25 Kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction