Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic 5 gallon Tight Head Pail Available on GSA/VA Contract for Federal Government customers only.

Seamless construction prevents leaks. Used for liquids.

Manufacturer:  Youngstown Barrel & Drum 2529

 View more versions of this product

Catalog No. 18-120-249


  Check Availability
Add to cart

Specifications

Description

Plastic 6 gal TH Round pail w/70mm cap. Seamless construction prevents leaks. UN rated 1H1/Y1.5/100. White. 11.37 in.dia x 17.62 in.H.
Description & Specifications

Specifications

Round tight head pail
44.75cm
28.87cm
6 gal.
70mm Screw Cap
11.37 in.
Plastic
250 class
4 lb.
Tight
17.62 in.
28.87cm
22.71L
UN 1H1/Y1.5/100
28.87cm
White
For transport of hazardous solids or liquids
1.81kg
70mm Screw Cap
6 gal., 22.71L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit