Learn More
Youngstown Barrel & Drum Plastic 5 gallon Tight Head Pail
Seamless construction prevents leaks. Used for liquids.
Manufacturer:  Youngstown Barrel & Drum 2532
 View more versions of this product
Catalog No. 18-120-250
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Plastic 6 Gal TH Rectangle pail w/70MM cap. Seamless construction prevents leaks. UN rated 3H1/Y1.8/100. White. 11.37 in. x 9.5 in. x 17.65 in.H.
Specifications
Specifications
UN 3H1/Y1.8/100
11.37 in.
For transport of hazardous solids or liquids
Tight
44.83cm
28.87cm
5 lb.
24.13cm
22.71L
Plastic
70mm Screw Cap
28.87cm
250 class
17.65 in.
70mm Screw Cap
Rectangular tight head pail
2.26kg
6 gal.
White