Learn More
Youngstown Barrel & Drum Ultra-Containment Berms™, Collapsible Wall Model Available on GSA/VA Contract for Federal Government customers only.
Economical design offers cost savings and secure containment
$10070.00
Specifications
Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Black
Material Copolymer 2000™ an Ethylene Copolymer, Copolymer 3000™, Polyurethane, XR5, PVC
Product Type Collapsible Wall Ultra-Containment Berm™
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Weight (English) Weight (Metric) Price Quantity & Availability  
19-339-014 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Youngstown Barrel & Drum
8402 UT
7405 gal. 28028L 420 lb. 191kg
Each for $10,070.00
Add to cart
Description
Description
Rugged design easily contains small and large spills
  • Heavy-duty copolymer 2000 construction with durable PVC sidewall assemblies for sidewall support
  • Raise or lower the sidewalls in seconds; simply swivel the feet of the PVC supports—vehicles and equipment can then roll over top the collapsed wall for convenient entry and exit
  • Fold up easily for compact storage and transport
  • Height: 12 in. (30.5cm)
Compliance
Meet EPA Containment Storage Regulation 40CFR264.175 with Collapsible Wall Model Containment Berm.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
40 CFR 264.175, SPCC, EPA
Black
Copolymer 2000™ an Ethylene Copolymer, Copolymer 3000™, Polyurethane, XR5, PVC
Collapsible Wall Ultra-Containment Berm™