Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-InLine Spill Decks™

Save valuable floor space

$1297.00 - $1416.00

Specifications

Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Yellow
Material Polyethylene
Product Type Ultra Inline Spill Deck
For Use With (Application) Reduces safety and handling concerns found with taller spill containment pallets
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Weight (English) Price Quantity & Availability  
19-339-041
View Documents
Youngstown Barrel & Drum
2333 UT
130 in. 7500 lb. 236 lb.
Each for $1,416.00
Add to cart
 
19-339-042
View Documents
Youngstown Barrel & Drum
2334 UT
156 in. 9000 lb. 291 lb.
Each for $1,297.00
Add to cart
 
Description

Description

Additional modules can be easily connected to an existing configuration as containment area storage needs expansion.

  • Bulkhead fittings lock connected modules together and allow spills to flow from one module to the next
  • 3-, 4- and 5-drum units utilize hidden containment bladder for additional sump capacity
  • Narrow 25.88 in. (66cm) depth allows drums to be stored in a single row along facility walls
  • Low-profile design reduces safety concerns
  • Pre-drilled models can be quickly assembled without tools

Compliance

Meet SPCC, EPA Container Storage Regulation 40 CFR 264.175 and UFC Spill Containment Regulations.

Specifications

Specifications

40 CFR 264.175, SPCC, EPA
Polyethylene
Reduces safety and handling concerns found with taller spill containment pallets
15cm
66cm
Yellow
Ultra Inline Spill Deck
5.75 in.
26 in.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit