Learn More
Youngstown Barrel & Drum Ultra-Oil Filter Boom, Skirted Model™
Stops oil spills at the surface and below
Manufacturer:  Youngstown Barrel & Drum 5241 UT
 View more versions of this product
Catalog No. 19-171-365
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Made with X-Tex material
  • Multiple booms can be connected for long, continuous runs
  • Easy deployment and retrieval
Specifications
Specifications
21 gal.
21 gal., 79.49L
For deeper water protection
X-Tex
12 lb.
79.49L
Gray
For use with ultra oil filter boom
Extension Skirt
5.44kg