Learn More
Youngstown Barrel & Drum Ultra-Safety Cabinet Bladder System™
Contains spills from drum strorage cabinets
$643.00 - $701.50
Specifications
Type Spill containment bladder system
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Weight (English) Price Quantity & Availability  
19-120-081
View Documents
Youngstown Barrel & Drum
2420 UT
Retrofit vertical one drum storage cabinet which fits under cabinet to capture spills 57 lb.
Each for $643.00
Add to cart
19-120-082
View Documents
Youngstown Barrel & Drum
2421 UT
Retrofit vertical two drum storage cabinet which fits under cabinet to capture spills 85 lb.
Each for $701.50
Add to cart
Description
Description
  • Retrofit all vertical drum storage cabinets to meet compliance requirements for containment of flammable and combustible liquids under EPA 40 CFR 264.175 and Uniform Fire Code Article 7901.8.5
  • To use raise existing drum cabinets 8 in. (20cm) from the floor, slide the bladder system in place, and lower the cabinet on top of the structural-steel frame
  • Significant leaks or spills unfurl the hidden containment bladder to provide complete spill containment; small spills are captured inside the polyethylene sump while significant leaks flow over the cabinet door sill and into the polyethylene sump
  • Heavy-duty, inner-steel framework keeps the cabinet stable, even in a fire
  • Low-profile design firmly positions drum cabinets only 6 1/4 in. (16cm) off the plant floor, so wastes can be poured into drum-top funnels at a convenient height with less risk for splashes into the eyes or face
Cabinet not included.
Specifications
Specifications
Spill containment bladder system