Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Deck™ P6 and P8

Low profile, modular spill decks for customized containment needs

$699.55 - $943.80

Specifications

Certifications/Compliance 40 CFR 264.175, SPCC, EPA
Color Yellow
Material Polyethylene
Type Ultra-Spill Deck
For Use With (Application) Perfect for satellite collection of hazardous wastes or storage of virgin chemicals
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Model Weight (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Model Weight (English) Price Quantity & Availability  
19-130-5061
View Documents Promotion Details
Youngstown Barrel & Drum
1175 UT
74.25 in. 9000 lb. 6-drum 86 lb.
Each for $699.55
Add to cart
 
19-130-5062
View Documents Promotion Details
Youngstown Barrel & Drum
1075 UT
98.5 in. 12000 lb. 8-drum 110 lb.
Each for $943.80
Add to cart
 
Description

Description

  • Perfect for satellite collection of hazardous wastes or storage of virgin chemicals
  • One-piece, molded sump with elevated 0.6 × 1.2m (2 ft. × 4 ft.) black grates
  • Polyethylene grates feature slip resistant “diamond plate” surface design
  • Deck holds up to 680kg (1500 lbs.) uniformly distributed load (UDL) per drum
  • Available in 6-drum and 8-drum models

Ultra Spill Deck P6

  • Containment capacity: 250L (66 gal.)
  • 1886 × 1270 × 133mm (74 × 50 × 5.2 in.)
  • Weight: 39kg (86 lbs.)
Ultra Spill Deck P8
  • Containment capacity: 284L (75 gal.)
  • 2502 × 1270 × 133mm (98.5 × 50 × 5.2 in.)
  • Weight: 50kg (110 lb.)

Order Info

Designed for use with Ultra-Spill Deck Loading Ramp (Cat. No. 19-058-033)

Compliance

Meets SPCC, EPA Container Storage Regulation 40 CFR 264.175 and United Fire Code Spill Containment Regulations

Specifications

Specifications

40 CFR 264.175, SPCC, EPA
Polyethylene
Perfect for satellite collection of hazardous wastes or storage of virgin chemicals
13.33 cm
50 in.
Yellow
Ultra-Spill Deck
5.25 in.
One-piece molded sump with elevated 0.5 ft. black polyethylene grates
127 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit