Learn More
Youngstown Barrel & Drum Ultra-Spill Decks™
Allow you to design and create a containment system that meets your specific needs
Manufacturer:  Youngstown Barrel & Drum 1090 UT
 View more versions of this product
Catalog No. 19-058-091
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Applications include hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage. Perfect for satellite collection of hazardous wastes.
  • Polyethylene construction meets a wide range of chemical compatibility needs
  • Bulkhead fittings lock connected modules together and allow spilled liquid to flow from one module to the next
  • Low profile (5.75 in.) height offers safe and convenient loading/unloading of drums
  • Easy to assemble, no tools required
Order Info
Optional loading ramp provides added drum handling safety.
Compliance
Meet EPA and Uniform Fire Code Spill Containment Regulations when assembled in configurations of six or more. Meet EPA Container Storage Regulation 40CFR264.175.
Specifications
Specifications
EPA, 40 CFR 264.175
Hazardous waste collection, storage of virgin chemicals, drum dispensing and battery storage
14.6cm
74.25 in.
50 in.
HDPE
Safety
74.25 x 50 x 5.75 in. (188.5 x 127 x 14.6cm)
5.75 in.
One 4 drum module, one 2 drum module and bulkhead fittings
188.5cm
127cm
Ultra-Spill Deck™ P6 System