Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Decks™

Allow you to design and create a containment system that meets your specific needs

$318.00 - $634.00

Specifications

Certifications/Compliance EPA, 40 CFR 264.175
Material HDPE
For Use With (Application) Hazardous waste collection, storage of virgin chemicals, drum dispensing and battery storage
Height (English) 5.75 in.
Height (Metric) 14.6cm
View More Specs

Products 5
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Weight (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Load Bearing Capacity (English) Weight (English) Width (English) Price Quantity & Availability  
19-058-075
Accessories View Documents
Youngstown Barrel & Drum
1086 UT
52 in. 3000 lb. 40 lb. 25.9 in.
Each for $513.50
Add to cart
 
19-058-043
Accessories View Documents
Youngstown Barrel & Drum
1321 UT
25.9 in. 1500 lb. 23 lb. 25.9 in.
Each for $349.50
Add to cart
 
19-058-074
View Documents
Youngstown Barrel & Drum
1072 UT
52 in. 6000 lb. 70 lb. 52 in.
Each for $634.00
Add to cart
 
19-058-033
View Documents
Youngstown Barrel & Drum
1089 UT
24 in. 600 lb. 17 lb. 32 in.
Each for $318.00
Add to cart
 
19-058-091
View Documents
Youngstown Barrel & Drum
1090 UT
74.25 in. 50 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Applications include hazardous waste collection, storage of virgin chemicals, drum dispensing, and battery storage. Perfect for satellite collection of hazardous wastes.

  • Polyethylene construction meets a wide range of chemical compatibility needs
  • Bulkhead fittings lock connected modules together and allow spilled liquid to flow from one module to the next
  • Low profile (5.75 in.) height offers safe and convenient loading/unloading of drums
  • Easy to assemble, no tools required

Order Info

Optional loading ramp provides added drum handling safety.

Compliance

Meet EPA and Uniform Fire Code Spill Containment Regulations when assembled in configurations of six or more. Meet EPA Container Storage Regulation 40CFR264.175.

Specifications

Specifications

EPA, 40 CFR 264.175
Hazardous waste collection, storage of virgin chemicals, drum dispensing and battery storage
14.6cm
HDPE
5.75 in.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit