Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Ultra-Spill Pallets Plus
Low-profile designonly 8.75 in. high (22.2cm)
$158.12 - $975.50
Specifications
Certifications/Compliance | SPCC, EPA, 40 CFR 264.175, UFC |
---|---|
Color | Yellow |
For Use With (Application) | Reduces the potential for hazardous fluids to splash into the eyes or face |
Catalog Number | Mfr. No. | Material | Product Type | Model | Weight (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Product Type | Model | Weight (English) | Price | Quantity & Availability | ||
19-130-4780
|
Youngstown Barrel & Drum
9631 UT |
Polyethylene | With drain | P4 | 140 lb. |
Each for $975.50
|
|
19-171-079
|
Youngstown Barrel & Drum
9606 UT |
Polyethylene | Without drain | P1 | 51 lb. |
Each for $381.00
|
|
19-130-5027
|
Youngstown Barrel & Drum
9610 UT |
Polyethylene | Without drain | P2 | 88 lb. |
Each for $595.00
|
|
18-938-502
|
Youngstown Barrel & Drum
9614 UT |
Vinyl | Pull Over Cover | Cover | 4 lb. |
Each for $158.12
|
|
Description
- Nearly 50% lower than standard spill pallets for safer and less strenuous handling
- Removable grates feature non-slip surface
- All-polyethylene construction
- Forkliftable
Order Info
Available in one-, two-, three- and four-drum configurations, with or without a drain
Compliance
Meet U.S. EPA container storage regulation 40 CFR 264.175, Uniform Fire Code spill containment regulations for use with drums and is SPCC compliant.
Specifications
SPCC, EPA, 40 CFR 264.175, UFC | |
Yellow | |
Reduces the potential for hazardous fluids to splash into the eyes or face |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title