Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Spill Deck™ Bladder Systems
SDP

Hidden containment bladder automatically unfurls to capture large spills or drum leaks

Specifications

Certifications/Compliance SPCC, EPA, 40 CFR 264.175, UFC
Color Yellow
Material Polyethylene
For Use With (Application) Used to capture large spills or drum leaks
Height (English) 5.75 in.
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Load Bearing Capacity (Metric) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Length (Metric) Load Bearing Capacity (Metric) Weight (Metric) Price Quantity & Availability  
19-058-005
SDP
View Documents
Youngstown Barrel & Drum
1320 UT
291 L 65.7 cm 680 kg 12.3 kg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-058-029
SDP
View Documents
Youngstown Barrel & Drum
2329 UT
333 L 132.08 cm 1360 kg 20 kg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Large catastrophic leaks flow into the bladder through a bulkhead fitting
  • Protective “trap door” opens automatically, allowing bladder to unfurl
  • Rugged, double polyethylene bladder allows 66 gal. (250L) of containment
  • Overspills and small leaks are captured in the the sump of the SpillDeck
  • Low profile 5.75 in. (14.6cm) deck height allows drums to be “jockeyed” onto SpillDeck Systems without the use of drum handling equipment

SpillDeck Bladder Systems require adequate aisle space to allow the bladder to unfurl properly.

Order Info

Available in one, two, and four-drum modules.

Compliance

Meets SPCC, EPA Container Storage Regulation 40 CFR 264.175 and UFC Spill Containment Regulations.

Specifications

Specifications

SPCC, EPA, 40 CFR 264.175, UFC
Polyethylene
5.75 in.
Grate, rugged double polyethylene bladder
77.47 cm
Yellow
Used to capture large spills or drum leaks
14.6 cm
30.5 in.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit