Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra Track Belts

Heavy-duty, reinforced PVC compound offers excellent oil/hydrocarbon resistance and long wear

$1034.43 - $1334.87

Specifications

Color Yellow/Black
Material PVC
Type Track Belt Set
For Use With (Application) Provide additional support between vehicles tires and containment berm surface
For Use With (Equipment) Economy containment berm
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
19-130-5005
View Documents
Youngstown Barrel & Drum
8343 UT
50 ft. 1270 cm
Pack of 2 for $1,034.43
Add to cart
 
19-130-5006
View Documents
Youngstown Barrel & Drum
8344 UT
60 ft. 1524 cm
Pack of 2 for $1,229.76
Add to cart
 
19-130-5007
View Documents
Youngstown Barrel & Drum
8345 UT
66 ft. 2011 cm
Pack of 2 for $1,334.87
Add to cart
 
Description

Description

Track belts are designed to be used with ultra-containment berms.

  • Material lies inside of containment berm
  • Runs the length of the berm's usable space
  • Acts as a buffer between vehicle tires and berm
  • All-polyester, single-ply interwoven carcass
  • Low stretch and good tear resistance
  • Color: Yellow and black

Specifications

Specifications

Yellow/Black
Track Belt Set
Economy containment berm
0.15 cm
76.2 cm
PVC
Provide additional support between vehicles tires and containment berm surface
0.062 in.
30 in.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit