Youngstown Barrel & Drum Pullover Covers for Ultra-Containment Berms™, Economy Models Available on GSA/VA Contract for Federal Government customers only.

For use with Ultra-Containment Berms, Economy Models (cat. no. 19171314)

Manufacturer: Youngstown Barrel & Drum Company 8333 UT

 View more versions of this product

Catalog No. 19-130-4720

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance 40 CFR 264.175, SPCC, EPA
    Color Yellow
    For Use With (Application) Protects drums and pallets from weather elements
    For Use With (Equipment) For 15 ft. x 66 ft. Ultra-spill containment berm™
    Product Type Pull Over Cover

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit