Youngstown Barrel & Drum Ultra-Containment Berm™, Foam Wall Models Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Youngstown Barrel & Drum Company 8387 UT

 View more versions of this product

Catalog No. 19-120-097

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 88 gal.
    Capacity (Metric) 333L
    Capacity 88 gal., 333L
    For Use With (Application) Capture nuisance leaks and spills from vehicles, oily equipment, tanks and drums
    Model Foam wall
    Weight (English) 83.77 lb.
    Weight (Metric) 38kg
    Material Copolymer 2000™
    Product Type Ultra-Containment Berm™ Foam Wall Model

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit