Youngstown Barrel & Drum Ultra-Containment Berms™ Stake Wall Models Available on GSA/VA Contract for Federal Government customers only.

Designed allowing the sidewalls to collapse in either direction and spring back automatically to their upright position

Manufacturer: Youngstown Barrel & Drum Company 8208 UT

 View more versions of this product

Catalog No. 19-339-015

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 179 gal.
    Capacity (Metric) 678L
    Capacity 179 gal., 678L
    Certifications/Compliance 40 CFR 264.175, SPCC, EPA
    For Use With (Application) Ideal for areas with temporary storage requirements
    For Use With (Equipment) Drums, tanks, equipment and vehicles
    Weight (English) 37 lb.
    Weight (Metric) 17kg
    Material XR5 an Ethylene Copolymer, Copolymer 3000™, Polyurethane
    Product Type Stake Wall Ultra-Containment Berm™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit