Learn More
YPEL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154031AP
Catalog No. 50-173-0202
Add to cart
Description
Description
15403-1-AP was raised against full length of YPEL3 protein (1-119 aa); it can recognize YPEL1-4.
YPEL3 (yippee-like 3) belongs to a family of five yippee-like proteins, all of which localize to the centrosome or mitotic spindle and are widely expressed in both adult and fetal tissue. This localization plus the fact that the family of human YPEL proteins share a high degree of sequence homology across species suggests that these proteins may have a conserved function involved in cell division. YPEL3 is a p53-regulated gene whose expression is induced by DNA damage and in turn induces cellular senescence. It appears to function as a tumor suppressor as it is downregulated in colon and breast tumors.
Specifications
Specifications
YPEL3
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61236, P61237
Protein yippee like 3, yippee like 3 (Drosophila), YPEL3
Rabbit
IgG
150 μL
-20°C
Primary
293491, 66090, 83719
Western Blot
Unconjugated
YPEL3
Liquid
Ypel3
YPEL3 Fusion Protein Ag7626
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat