Learn More
YSI™ 60510 Pro Series ISE Cable
The 60510 is a 1-port cable for use with the Professional Plus instrument. The port requires a ISE probe.
$517.50 - $621.00
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
15-176-203
|
YSI™
6051010 |
393.7 in. | 10 m |
Each for $517.50
|
|
||||
15-177-468
|
YSI™
6051020 |
787.4 in. | 20 m |
Each for $621.00
|
|
||||
Description
Conductivity Probes and Multiparameter Cables. Order other probes separately.
- The 60510 ISE (pH, ORP, pH/ORP, ammonium, nitrate, chloride) cables ONLY work with the YSI Pro Plus handheld Any of the ISE sensors can be connected to this cable at any time
Specifications
Cable management kit |