Learn More
YSI™ 6052030 Pro Series DO/Conductivity Cables
Manufacturer:  YSI™ 60520304
 View more versions of this product
Catalog No. 15-177-490
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Conductivity, DO, Temp
157.4 in.
Built-in conductivity sensor and temperature/DO sensor port
Professional Plus and Pro2030 DO/conductivity meter
4m