Learn More
YSI™ Colorimeter Reagents - Ammonium
Manufacturer:  YSI™ 251997Y
 View more versions of this product
Catalog No. 15-177-801
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
50/Pk.
0.4 to 50mg/L
Used to measure ammonium
50.0
EPA
Powder
Photoflex colorimeter
20°C