Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Colorimeter Reagents - Ammonium

$33.90 - $91.70

Specifications

Format Powder
For Use With (Equipment) Photoflex colorimeter
View More Specs

Products 7
Catalog Number Mfr. No. For Use With (Application) Quantity No. of Tests Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Quantity No. of Tests Price Quantity & Availability  
15-177-751
View Documents
YSI™
251408Y
Used to measure ammonium 100/Pk. 100
Pack of 100 for $121.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-763
View Documents
YSI™
251997RP1Y
Used to measure ammonium, ammonia cyanurate replacement pack for ammonium 50/Pk. 50
Pack of 50 for $22.95
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-764
View Documents
YSI™
251408RP2Y
Used to measure ammonium, ammonia salicylate replacement pack for ammonium 100/Pk. 100
Pack of 100 for $68.10
Add to cart
 
15-177-766
View Documents
YSI™
251408RP1Y
Used to measure ammonium, ammonia powder replacement pack for ammonium 100/Pk. 100
Pack of 100 for $44.75
Add to cart
 
15-177-765
View Documents
YSI™
251997RP2Y
Used to measure ammonium, ammonia salicylate replacement pack for ammonium 50/Pk. 50
Pack of 50 for $33.90
Add to cart
 
15-177-800
View Documents
YSI™
251998Y
Used to measure ammonium 50/Pk. 50
Pack of 25 for $91.70
Add to cart
 
15-177-801
View Documents
YSI™
251997Y
Used to measure ammonium 50/Pk. 50
Pack of 25 for $102.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Specifications

Specifications

Powder
Photoflex colorimeter
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit