Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
Supplier: YSI™ 251414LP1Y
Description
- DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
DPD | |
Powder | |
Photoflex or 900 colorimeter | |
0.338 oz. |
10 mL | |
Used to measure total chlorine | |
0.02 to 2mg/L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction