Learn More
YSI™ Colorimeter Reagents - Chlorine
Chlorine reagents for various colorimeters
$16.36 - $204.00
Specifications
Format Powder
View More Specs
Products 15
Catalog Number Mfr. No. Description Range For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Description Range For Use With (Application) Price Quantity & Availability  
15-177-729
View Documents
YSI™
251401Y
DPD 0.02 to 2mg/L Used to measure free chlorine
Pack of 100 for $24.65
Add to cart
15-177-731
View Documents
YSI™
251415Y
DPD 0.05 to 5mg/L Used to measure total chlorine
Pack of 100 for $26.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-730
View Documents
YSI™
251402Y
DPD 0.05 to 5mg/L Used to measure free chlorine
Pack of 100 for $25.15
Add to cart
15-177-733
View Documents
YSI™
251401LP1Y
DPD 0.02 to 2mg/L Used to measure free chlorine
Pack of 1000 for $200.00
Add to cart
15-177-735
View Documents
YSI™
251398Y
DPD 0 to 3.5mg/L Used to measure free chlorine
Pack of 100 for $19.95
Add to cart
15-177-736
View Documents
YSI™
251398LPY
DPD 0 to 3.5mg/L Used to measure free chlorine
Pack of 1000 for $159.30
Add to cart
15-177-737
View Documents
YSI™
251414LP1Y
DPD 0.02 to 2mg/L Used to measure total chlorine
Pack of 1000 for $181.65
Add to cart
15-177-734
View Documents
YSI™
251402LP1Y
DPD 0.05 to 5mg/L Used to measure free chlorine
Pack of 1000 for $198.60
Add to cart
15-177-738
View Documents
YSI™
251415LP1Y
DPD 0.05 to 5mg/L Used to measure total chlorine
Pack of 1000 for $204.00
Add to cart
15-177-739
View Documents
YSI™
251397Y
DPD 0 to 3.5mg/L Used to measure total chlorine
Pack of 100 for $21.25
Add to cart
15-177-740
View Documents
YSI™
251397LPY
DPD 0 to 3.5mg/L Used to measure total chlorine
Pack of 1000 for $158.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-807
View Documents
YSI™
251600Y
DPD No. 1 LR 0.01 to 6mg/L Used to measure free chlorine
Pack of 100 for $17.44
Add to cart
15-177-808
View Documents
YSI™
251601Y
DPD No. 3 LR 0.01 to 6mg/L Used to measure free chlorine
Pack of 100 for $17.44
Add to cart
15-177-809
View Documents
YSI™
251602Y
DPD No. 1 HR 0.1 to 10mg/L Used to measure free chlorine
Pack of 100 for $17.44
Add to cart
15-177-810
View Documents
YSI™
251603Y
DPD No. 3 HR 0.1 to 10mg/L Used to measure free chlorine
Pack of 100 for $16.36
Add to cart
Description
Description
  • DPD reagents for measuring chlorine
Compliance
EPA Approved Reagents: Cl2-1 TP, Cl2T1TT, Cl2T2 TT, Cl2T3 TT, Cl2T4 TT
Specifications
Specifications
Powder