YSI™ Colorimeter Reagents - Chlorine

Chlorine reagents for various colorimeters

Manufacturer: YSI™ 251397Y

 View more versions of this product

Catalog No. 15-177-739

  Check Availability
Add to Cart

    Specifications

    Format Powder
    For Use With (Application) Used to measure total chlorine
    For Use With (Equipment) Used with other manufacturers colorimeter
    pH Range 6.3
    Storage Requirements 20°C
    Volume (English) 0.169 oz.
    Volume (Metric) 5mL
    Color White
    Range 0 to 3.5mg/L

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit