Learn More
YSI™ Colorimeter Reagents - COD
Manufacturer:  YSI™ 251991Y
 View more versions of this product
Catalog No. 15-177-795
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
25/Pk.
COD2 TC (Mid Range)
Used to measure COD
EPA
20 to 1500mg/L
Photoflex colorimeter