YSI™ Colorimeter Reagents - COD
Manufacturer: YSI™ 251992Y
 View more versions of this product
Catalog No. 15-177-796
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Used to measure COD
    For Use With (Equipment) Photoflex colorimeter
    Quantity 25/Pk.
    Certifications/Compliance EPA
    Product Type COD3 TC (High Range)
    Range 200 to 15000mg/L
  • Description & Specifications
Related Products
Product Content Feedback