Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Colorimeter Reagents - COD
$65.50 - $442.00
Specifications
For Use With (Application) | Used to measure COD |
---|---|
For Use With (Equipment) | Photoflex colorimeter |
Catalog Number | Mfr. No. | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Range | Price | Quantity & Availability | ||||
15-177-791
|
YSI™
251984Y |
COD3-F TC (High Range), Hg free | 0 to 1500mg/L |
Pack of 25 for $65.50
|
|
||||
15-177-789
|
YSI™
251985Y |
COD2-F TC (Mid Range), Hg free | 0 to 1500mg/L |
Pack of 25 for $65.50
|
|
||||
15-177-792
|
YSI™
251992LPY |
COD3 TC (High Range) | 200 to 15000mg/L |
Pack of 150 for $432.00
|
|
||||
15-177-790
|
YSI™
251991LPY |
COD2 TC (Mid Range) | 20 to 1500mg/L |
Pack of 150 for $442.00
|
|
||||
15-177-794
|
YSI™
251990LPY |
COD1 TC (Low Range) | 3 to 150mg/L |
Pack of 150 for $432.00
|
|
||||
15-177-793
|
YSI™
251986Y |
COD1-F TC (Low Range), Hg free | 0 to 150mg/L |
Pack of 25 for $65.50
|
|
||||
15-177-795
|
YSI™
251991Y |
COD2 TC (Mid Range) | 20 to 1500mg/L |
Pack of 25 for $65.50
|
|
||||
15-177-796
|
YSI™
251992Y |
COD3 TC (High Range) | 200 to 15000mg/L |
Pack of 25 for $65.50
|
|
||||
15-177-797
|
YSI™
251990Y |
COD1 TC (Low Range) | 3 to 150mg/L |
Pack of 25 for $66.90
|
|
||||
Specifications
Used to measure COD | |
Photoflex colorimeter |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title