YSI™ Colorimeter Reagents - COD
Manufacturer: YSI™ 251985Y
 View more versions of this product
Catalog No. 15-177-789
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Used to measure COD
    For Use With (Equipment) Photoflex colorimeter
    Quantity 25/Pk.
    Product Type COD2-F TC (Mid Range), Hg free
    Range 0 to 1500mg/L
  • Description & Specifications
Related Products
Product Content Feedback