Learn More
YSI™ Colorimeter Reagents - Iron
Manufacturer:  YSI™ 251405Y
 View more versions of this product
Catalog No. 15-177-742
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
100/Pk.
EPA
0.02 to 3mg/L
Used to measure total iron
5.6
White
Iron Fe-2 TP, Total
Powder
Photoflex colorimeter
20°C