Learn More
YSI™ Colorimeter Reagents - Iron
Manufacturer:  YSI™ 251405LPY
 View more versions of this product
Catalog No. 15-177-743
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1000/Pk.
Iron Fe-2 TP, Total
Powder
Photoflex colorimeter
20°C
White
0.02 to 3mg/L
Used to measure total iron
5.6