Learn More
YSI™ Colorimeter Reagents - Manganese
Manufacturer:  YSI™ 251417RP6Y
 View more versions of this product
Catalog No. 15-177-823
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Manganese Mn-2 RP6
PAN indicator replacement pack for manganese Mn-2 TP
3.38 oz.
Liquid
Photoflex colorimeter
100mL