Learn More
YSI™ Colorimeter Reagents - Manganese
$14.17 - $69.90
Specifications
For Use With (Equipment) Photoflex colorimeter
View More Specs
Products 13
Catalog Number Mfr. No. Type Format For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Type Format For Use With (Application) Price Quantity & Availability  
15-177-745
View Documents Promotion Details
YSI™
251417Y
Manganese Mn-2 TP (Low Range) Powder Used to measure low range manganese
Pack of 50 for $48.30
Add to cart
15-177-744
View Documents Promotion Details
YSI™
251406Y
Manganese Mn-1 TP (High Range) Powder Used to measure high range manganese
Pack of 100 for $54.60
Add to cart
15-177-767
View Documents Promotion Details
YSI™
251417RP2Y
Manganese Mn-2 RP2 Powder Ascorbic acid replacement pack for manganese Mn-2 TP se
Pack of 100 for $22.95
Add to cart
15-177-771
View Documents Promotion Details
YSI™
251406RP1Y
Manganese Mn-1 RP1 Powder Replacement pack for manganese Mn-1 TP
Pack of 100 for $36.05
Add to cart
15-177-787
View Documents Promotion Details
YSI™
251406RP2Y
Manganese Mn-1 RP2 Powder Sodium periodate replacement pack for manganese
Pack of 100 for $28.40
Add to cart
15-177-812
View Documents Promotion Details
YSI™
251417A1
Manganese Mn-2 A1 Liquid For use with manganese Mn-2 TP set
Each for $14.17
Add to cart
15-177-813
View Documents Promotion Details
YSI™
251417A2
Manganese Mn-2 A2 Liquid For use with manganese Mn-2 TP set
Each for $69.90
Add to cart
15-177-814
View Documents Promotion Details
YSI™
251417RP4Y
Manganese Mn-2 RP4 Liquid Replacement pack for manganese Mn-2 TP
Each for $25.15
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-815
View Documents Promotion Details
YSI™
251417RP5Y
Manganese Mn-2 RP5 Liquid Alkaline cyanide replacement pack for manganese Mn-2 TP set
Each for $63.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-816
View Documents Promotion Details
YSI™
251417RP1Y
Manganese Mn-2 RP1 Liquid Alkaline cyanide replacement pack for manganese Mn-2 TP
Pack of 50 for $20.75
N/A
Due to product restrictions, please to purchase or view availability for this product.
15-177-821
View Documents Promotion Details
YSI™
251417RP7Y
Manganese Mn-2 RP7 Liquid PAN indicator replacement pack for manganese Mn-2 TP
Each for $63.00
Add to cart
15-177-822
View Documents Promotion Details
YSI™
251417RP3Y
Manganese Mn-2 RP3 Liquid PAN indicator replacement pack for manganese Mn-2 TP
Each for $17.44
Add to cart
15-177-823
View Documents Promotion Details
YSI™
251417RP6Y
Manganese Mn-2 RP6 Liquid PAN indicator replacement pack for manganese Mn-2 TP
Each for $26.20
Add to cart
Description
Description
Specifications
Specifications
Photoflex colorimeter