Learn More
YSI™ Colorimeter Reagents - Nitrogen
Manufacturer:  YSI™ 251996Y
 View more versions of this product
Catalog No. 15-177-802
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
50/Pk.
10 to 150mg/L
Used to measure total nitrogen
25.0
Nitrogen Ntot2 TC (High range)
Vial
Photoflex colorimeter