Learn More
YSI™ Colorimeter Reagents - Phosphate
$13.65 - $266.50
Specifications
For Use With (Equipment) Photoflex colorimeter
View More Specs
Products 9
Catalog Number Mfr. No. Type Format For Use With (Application) No. of Tests Price Quantity & Availability  
Catalog Number Mfr. No. Type Format For Use With (Application) No. of Tests Price Quantity & Availability  
15-177-753
View Documents Promotion Details
YSI™
251410Y
Phosphate PO4-1 TP Powder Used to measure orthophosphate 100
Pack of 100 for $40.85
Add to cart
15-177-754
View Documents Promotion Details
YSI™
251410LPY
Phosphate PO4-1 TP Powder Used to measure orthophosphate 1000
Pack of 1000 for $266.50
Add to cart
15-177-779
View Documents Promotion Details
YSI™
251987RP1Y
Phosphate PO4-4 RP1 (replacement pack) Powder Phosphate 3 replacement pack for Phosphate PO4-4 TC 50
Pack of 50 for $17.44
Add to cart
15-177-780
View Documents Promotion Details
YSI™
251987RP2Y
Phosphate PO4-4 RP2 (replacement pack) Powder Potassium persulfate replacement pack for Phosphate PO4-4 TC 50
Pack of 50 for $18.54
Add to cart
15-177-804
View Documents Promotion Details
YSI™
251987Y
Phosphate PO4-4 TC Vial Used to measure acid hydrolyzable phosphorus 50
Pack of 50 for $65.50
Add to cart
15-177-805
View Documents Promotion Details
YSI™
251989Y
Phosphate PO4-2 TP Vial Used to measure reactive orthophosphate 100
Pack of 50 for $44.75
Add to cart
15-177-806
View Documents Promotion Details
YSI™
251988Y
Phosphate PO4-3 TC Vial Used to measure total phosphorus 50
Pack of 50 for $68.40
Add to cart
15-177-824
View Documents Promotion Details
YSI™
251987RP4Y
Phosphate PO4-4 RP4 (replacement pack) Liquid Sodium hydroxide (1N) replacement pack for Phosphate PO4-4 TC 50
Each for $13.65
Add to cart
15-177-825
View Documents Promotion Details
YSI™
251987RP3Y
Phosphate PO4-4 RP3 (replacement pack) Liquid Sodium hydroxide (1.54N) replacement pack for Phosphate PO4-4 TC 50
Each for $13.65
Add to cart
Description
Description
Specifications
Specifications
Photoflex colorimeter