Learn More
YSI™ Colorimeter Reagents - Sulfate
Manufacturer:  YSI™ 251423Y
 View more versions of this product
Catalog No. 15-177-761
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
EPA
Used to measure sulfate
0.84 oz.
2 to 70mg/L
Photoflex colorimeter
25mL