Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
YSI™ Dip-Type Glass Conductivity Cells
Manufacturer:  YSI™ 603401
 View more versions of this product
Catalog No. 09-324-31
Specifications
Specifications
1cm-1
1 in.
3100 and 3200 Meter
48 in.
1.2m
Conductivity
Banana
25mm
7.5 in.
482.6 mm
Pyrex 7740 Glass
No
Documents
Accessories