Learn More
YSI™ Dip-Type Glass Conductivity Cells
Manufacturer:  YSI™ 603402
 View more versions of this product
Catalog No. 09-324-32
Add to cart
Specifications
Specifications
0.1cm-1
1 in.
3200 Meter
48 in.
1.2m
Conductivity
Banana
25mm
6.25 in.
406.4mm
Pyrex 7740 Glass
No
Documents
Accessories