Learn More
YSI™ Dip-Type Plastic Conductivity Cells
YSI dip-type ABS-plastic conductivity cell for use with YSI conductivity meters (sold separately).
Manufacturer:  YSI™ 099820
 View more versions of this product
Catalog No. 09-324-28
Add to cart
Description
Description
Dip cell requires cell adapter (sold separately).
Specifications
Specifications
1cm-1
Banana
13mm
5.74 in.
146mm
ABS
No
Conductivity meters, also known as electrical conductivity (EC) meters, are used in research laboratories and a variety of industries, such as water-quality testing, waste-water treatment, hydroponics, agriculture, aquaculture, and cosmetics manufacturing.
0.5 in.
3100 and 3200 Meter
48 in.
1.2m
Conductivity
Documents
Accessories