Learn More
YSI™ Field Dissolved Oxygen Probes Products for Science Education
Versatile, easy-to-use polarographic electrode that uses detachable cables.
Manufacturer:  YSI™ 057087/EMD
Catalog No. S60036
Add to cart
Description
Description
  • Rugged field probe with built-in weight
  • Detachable cable assemblies
  • Automatic pressure compensation
  • Compatible with the YSI 52 or 58 Dissolved Oxygen Meters
Specifications
Specifications
1 Year
Field use
DO polarographic probe
59° to 95°F
±1mg/L
YSI portable model 52 and 58 or YSI 5000 and 5100 benchtop meter
90% in 30 sec.
15° to 35°C