Learn More
YSI™ Fill Conductivity Cell
Supplier: YSI™ 603254
Specifications
1 cm-1 | |
7-pin Mini-DIN | |
19.05 mm | |
48 in. | |
1.2 m | |
Conductivity |
3200 and 3400 Meter | |
0.75 in. | |
5.25 in. | |
135 mm | |
Pyrex 7740 Glass | |
Yes |