Learn More
YSI™ MultiLab Line Benchtop Laboratory Instruments
For accurate pH, ORP, conductivity and BOD measurements in lab
Manufacturer:  YSI™ 1FD460Y
 View more versions of this product
Catalog No. 09-324-559
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Parameters measured: pH, ORP (mV), DO%, DO mg/L (BOD probe), Partical pressure, Conductivity, Resistivity, Salinity, TDS, Temperature
  • Large, easy to read graphic display
  • Intelligent, digital sensors with plug and play
  • Mini USB connectivity to manage data
  • Temperature compensation: yes (except ORP)
Certifications
CE marked cETLus
Compliance
GLP/AQA compliant
Warranty and Services
Three years
Specifications
Specifications
3 Years
DO and conductivity = 1, pH = 1 to 5
Color Graphic Display
CE, cETLus
1.5V
2
Mini USB, USB-A