Learn More
YSI™ MultiLab Line BOD Laboratory Kits
Kit for BOD calculation
$2950.00
Specifications
Channels 1
View More Specs
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
09-324-565
View Documents Promotion Details
YSI™
626570
4010-1 One channel benchtop instrument, optical BOD probe
Each for $2,950.00
Add to cart
Description
Description
BOD Lab Kit Includes
  • 4010-1 One Channel benchtop instrument
  • Optical BOD probe
Advanced BOD Lab Kit Includes
  • 4010-1 One Channel benchtop instrument
  • Optical BOD probe
  • BOD Analyst Pro desktop software
Specifications
Specifications
1