Learn More
YSI™ Pro Plus Handheld Multiparameter Instrument
Supplier: YSI™ 603180
Specifications
0.001mS/cm | |
±20mV in redox standards | |
±1.0% of reading | |
RoHS, CE, WEEE, C-Tick, VCCI, FCC, IP-67, 1-meter drop test, Assembled in USA | |
4m Cable with Built-In Conductivity and Polarographic DO Probe | |
32°F to 104°F | |
3.25 in. | |
475 g |
0 to 200% | |
±0.2 | |
1, 2, 3, 4, 5, or 6 point | |
0°C to 40°C | |
±0.35°C | |
3 years instrument, 2 years field cables | |
1.05 lb. |